Assay Details
Target Gene Details
Entrez Gene ID: | 3938 |
Gene Name: | lactase |
Gene Aliases: |
LAC, LPH, LPH1 |
Location: |
Chr.2:135787840-135837200 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCT | NM_002299.3 | NP_002290.2 | ||
| X07994.1 | CAA30801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592582 | Chr.2:135755875 - 135805055 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map