Assay Details
Target Gene Details
Entrez Gene ID: | 6303 |
Gene Name: | spermidine/spermine N1-acetyltransferase 1 |
Gene Aliases: |
DC21, KFSD, KFSDX, SAT, SSAT, SSAT-1 |
Location: |
Chr.X:23783158-23786223 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAT1 | NM_002970.3 | NP_002961.1 | ||
| NR_027783.2 | ||||
| AF251292.1 | AAG44592.1 | |||
| AK310078.1 | 3 | 367 | ||
| AK310094.1 | 2 | 651 | ||
| AK312162.1 | ||||
| AK315888.1 | 3 | 292 | ||
| AL050290.1 | ||||
| AY841998.1 | AAW03315.1 | |||
| BC002503.2 | AAH02503.1 | |||
| BC008424.1 | AAH08424.1 | |||
| BT006825.1 | AAP35471.1 | |||
| CD516700.1 | ||||
| CD638929.1 | ||||
| CN271682.1 | ||||
| CR450294.1 | CAG29290.1 | |||
| M77693.1 | AAA60573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470346 | Chr.X:23738748 - 23786754 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map