Assay Details
Target Gene Details
Entrez Gene ID: | 84681 |
Gene Name: | histidine triad nucleotide binding protein 2 |
Gene Aliases: |
HIT-17 |
Location: |
Chr.9:35812960-35815045 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HINT2 | NM_032593.2 | NP_115982.1 | ||
| AF085236.1 | AAL40394.1 | |||
| AF356515.1 | AAK37562.1 | |||
| AF356875.1 | AAM00221.1 | |||
| AK222582.1 | BAD96302.1 | |||
| AY033094.1 | AAK53455.1 | |||
| BC047737.2 | AAH47737.1 | |||
| BI759951.1 | ||||
| BM764627.1 |
More Information
Set Membership: |
|

Genomic Map