Assay Details
Target Gene Details
Entrez Gene ID: | 83733 |
Gene Name: | solute carrier family 25 member 18 |
Gene Aliases: |
GC2 |
Location: |
Chr.22:17562470-17590890 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A18 | NM_001303484.1 | 1 | 216 | NP_001290413.1 |
| NM_031481.2 | 1 | 216 | NP_113669.1 | |
| XM_011546149.2 | 1 | 1156 | XP_011544451.1 | |
| XM_011546150.2 | 1 | 1158 | XP_011544452.1 | |
| XM_011546153.2 | 1 | 1158 | XP_011544455.1 | |
| XM_017028969.1 | 1 | 1152 | XP_016884458.1 | |
| XM_017028970.1 | 1 | 1152 | XP_016884459.1 | |
| XM_017028971.1 | 1 | 1152 | XP_016884460.1 | |
| XM_017028972.1 | 1 | 1152 | XP_016884461.1 | |
| XM_017028973.1 | 1 | 1152 | XP_016884462.1 | |
| XM_017028974.1 | 1 | 1152 | XP_016884463.1 | |
| XM_017028975.1 | 1 | 1152 | XP_016884464.1 | |
| AJ428203.1 | 1 | 172 | CAD21008.1 | |
| AK095706.1 | 1 | 66 | ||
| AK308974.1 | 1 | 170 | ||
| AK309110.1 | 1 | 63 | ||
| AY008285.1 | 1 | 172 | AAG22855.1 | |
| BC031644.1 | 1 | 171 | AAH31644.1 | |
| DA354203.1 | 1 | 216 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss |
|
| nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain |
|
| nsv1065055 | Chr.22:17559085 - 17632626 on Build GRCh38 | Gain |
|
| nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss |
|
| nsv510802 | Chr.22:17550100 - 17604891 on Build GRCh38 | Deletion |
|
| nsv3553 | Chr.22:17553601 - 17608096 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map