Assay Details
Target Gene Details
Entrez Gene ID: | 124446 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
IGFBP-3R |
Location: |
Chr.16:29962030-29973052 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM219 | NM_001083613.1 | NP_001077082.1 | ||
| NM_194280.3 | NP_919256.1 | |||
| XM_005255123.1 | XP_005255180.1 | |||
| BC017488.1 | AAH17488.1 | |||
| BC050051.1 | AAH50051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29470 | Chr.16:29972742 - 29973532 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map