Assay Details
Target Gene Details
Entrez Gene ID: | 147429 |
Gene Name: | AQP4 antisense RNA 1 |
Gene Aliases: |
C18orf16, CHST9-AS1 |
Location: |
Chr.18:26865308-26935946 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AQP4-AS1 | NR_026908.1 | |||
| AK055069.1 | BAB70850.1 |
Target Gene Details
Entrez Gene ID: | 83539 |
Gene Name: | carbohydrate sulfotransferase 9 |
Gene Aliases: |
GALNAC4ST-2, GalNAc4ST2 |
Location: |
Chr.18:26908754-27185364 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHST9 | NM_001256316.1 | 5 | 1183 | NP_001243245.1 |
| NM_031422.5 | 6 | 1221 | NP_113610.2 | |
| XM_005258362.3 | 2 | 769 | XP_005258419.1 | |
| XM_017026033.1 | 5 | 1120 | XP_016881522.1 | |
| XM_017026034.1 | 7 | 1344 | XP_016881523.1 | |
| AF239821.1 | 6 | 1098 | AAK01862.1 | |
| AF332472.1 | 5 | 1012 | AAK30369.1 | |
| AF332473.1 | 3 | 779 | AAK30370.1 | |
| AK093349.1 | 5 | 1120 | BAC04141.1 | |
| AY358488.1 | 6 | 1200 | AAQ88852.1 | |
| BC025764.1 | 6 | 1165 | AAH25764.2 | |
| KU178792.1 | 5 | 907 | ||
| KU178793.1 | 4 | 869 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761995 | Chr.18:26634322 - 26922606 on Build GRCh38 | Loss |
|
| nsv833600 | Chr.18:26893930 - 27101055 on Build GRCh38 | Gain |
|
| esv3893079 | Chr.18:26912135 - 27610887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map