Assay Details
Target Gene Details
Entrez Gene ID: | 10519 |
Gene Name: | calcium and integrin binding 1 |
Gene Aliases: |
CIB, CIBP, KIP1, PRKDCIP, SIP2-28 |
Location: |
Chr.15:90230245-90265759 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB1 | NR_102428.1 | |||
| BG759961.1 |
Target Gene Details
Entrez Gene ID: | 440307 |
Gene Name: | tubulin tyrosine ligase like 13, pseudogene |
Gene Aliases: |
TTLL13 |
Location: |
Chr.15:90249532-90259088 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL13P | NR_104604.1 | |||
| AK127965.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss |
|
| nsv1052652 | Chr.15:90101186 - 90298961 on Build GRCh38 | Gain |
|
| esv3637218 | Chr.15:90252532 - 90266165 on Build GRCh38 | Gain |
|
| esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain |
|
| esv2751541 | Chr.15:90245652 - 90457213 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map