Assay Details
Target Gene Details
Entrez Gene ID: | 29070 |
Gene Name: | coiled-coil domain containing 113 |
Gene Aliases: |
HSPC065 |
Location: |
Chr.16:58196863-58283836 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC113 | NM_001142302.1 | 8 | 4654 | NP_001135774.1 |
| NM_014157.3 | 9 | 4816 | NP_054876.2 | |
| XM_011523038.2 | 9 | 4671 | XP_011521340.1 | |
| XM_011523039.2 | 11 | 5564 | XP_011521341.1 | |
| XM_011523040.2 | 9 | 5089 | XP_011521342.1 | |
| XM_011523041.2 | 9 | 4657 | XP_011521343.1 | |
| XM_017023176.1 | 10 | 4979 | XP_016878665.1 | |
| XM_017023177.1 | 8 | 4673 | XP_016878666.1 | |
| AK027208.1 | 1 | 1951 | BAB15692.1 | |
| AL136785.1 | 9 | 4793 | CAB66719.1 |
Target Gene Details
Entrez Gene ID: | 221191 |
Gene Name: | protease, serine 54 |
Gene Aliases: |
CT67, KLKBL4 |
Location: |
Chr.16:58279997-58295047 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS54 | NM_001080492.1 | NP_001073961.1 | ||
| NM_001305173.1 | NP_001292102.1 | |||
| NM_001305174.1 | NP_001292103.1 | |||
| AK058068.1 | BAB71649.1 | |||
| AK302175.1 | ||||
| AL137484.1 | CAB70765.1 | |||
| BC057843.1 |
More Information
Set Membership: |
|

Genomic Map