Assay Details
Target Gene Details
Entrez Gene ID: | 9507 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 4 |
Gene Aliases: |
ADAMTS-2, ADAMTS-4, ADMP-1 |
Location: |
Chr.1:161189725-161199080 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS4 | NM_001320336.1 | 1 | 801 | NP_001307265.1 |
| NM_005099.5 | 1 | 801 | NP_005090.3 | |
| AB014588.1 | 1 | 775 | ||
| AF148213.1 | 1 | 781 | AAD41494.1 | |
| AK291573.1 | 1 | 800 | ||
| AY358886.1 | 1 | 793 | AAQ89245.1 | |
| BC063293.1 | 1 | 800 | AAH63293.1 |
Target Gene Details
Entrez Gene ID: | 4720 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S2 |
Gene Aliases: |
CI-49 |
Location: |
Chr.1:161197377-161214395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS2 | XM_005245208.2 | XP_005245265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32799 | Chr.1:161192048 - 161201730 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map