Assay Details
Target Gene Details
Entrez Gene ID: | 90634 |
Gene Name: | NEDD4 binding protein 2 like 1 |
Gene Aliases: |
CG018 |
Location: |
Chr.13:32400723-32429649 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
N4BP2L1 | NM_001079691.1 | 5 | 716 | NP_001073159.1 |
NM_001286459.1 | 5 | 724 | NP_001273388.1 | |
NM_001286460.1 | NP_001273389.1 | |||
NM_001286461.1 | NP_001273390.1 | |||
NM_052818.2 | 5 | 724 | NP_438169.2 | |
XM_006719890.3 | 6 | 1086 | XP_006719953.1 | |
XM_011535301.2 | 6 | 877 | XP_011533603.1 | |
XM_011535302.2 | 8 | 1158 | XP_011533604.1 | |
XM_011535303.2 | 5 | 1287 | XP_011533605.1 | |
XM_011535304.2 | 8 | 1135 | XP_011533606.1 | |
XM_017020836.1 | 6 | 2277 | XP_016876325.1 | |
XM_017020837.1 | 4 | 887 | XP_016876326.1 | |
XM_017020838.1 | 8 | 1218 | XP_016876327.1 | |
XM_017020840.1 | 8 | 1133 | XP_016876329.1 | |
XM_017020841.1 | 8 | 1226 | XP_016876330.1 | |
XM_017020842.1 | 9 | 1264 | XP_016876331.1 | |
AI916535.1 | ||||
AL049785.1 | 5 | 680 | CAB42443.1 | |
AL049786.1 | 5 | 680 | CAB42444.1 | |
AL049787.1 | 5 | 672 | CAB42445.1 | |
AL049788.1 | 7 | 738 | CAB42446.1 | |
BC022188.2 | 5 | 662 | AAH22188.2 | |
BC125050.1 | 4 | 393 | ||
BF448179.1 | ||||
BI551105.1 | ||||
BX648509.1 | 4 | 3653 | ||
U50527.1 | 5 | 667 | ||
U57962.1 | 1 | 1070 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832576 | Chr.13:32322994 - 32502851 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv976257 | Chr.13:32401379 - 32404465 on Build GRCh38 | Duplication |
![]() |
