Assay Details
Target Gene Details
Entrez Gene ID: | 113 |
Gene Name: | adenylate cyclase 7 |
Gene Aliases: |
AC7 |
Location: |
Chr.16:50246158-50318135 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY7 | NM_001114.4 | 25 | 5833 | NP_001105.1 |
| XM_011522835.2 | 26 | 5952 | XP_011521137.1 | |
| XM_011522836.2 | 26 | 5968 | XP_011521138.1 | |
| XM_011522837.2 | 26 | 5897 | XP_011521139.1 | |
| XM_011522838.2 | 26 | 6101 | XP_011521140.1 | |
| XM_011522839.2 | 26 | 5928 | XP_011521141.1 | |
| XM_011522840.2 | 26 | 5935 | XP_011521142.1 | |
| XM_011522842.2 | 26 | 5998 | XP_011521144.1 | |
| XM_017022895.1 | 26 | 5950 | XP_016878384.1 | |
| XM_017022896.1 | 23 | 5114 | XP_016878385.1 | |
| XM_017022897.1 | 23 | 5133 | XP_016878386.1 | |
| AA740624.1 | 1 | 322 | ||
| BM543987.1 | 1 | 74 | ||
| BU631614.1 | 1 | 324 | ||
| D25538.1 | 25 | 5830 |
Target Gene Details
Entrez Gene ID: | 29117 |
Gene Name: | bromodomain containing 7 |
Gene Aliases: |
BP75, CELTIX1, NAG4 |
Location: |
Chr.16:50317563-50368969 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRD7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1805 | Chr.16:50307115 - 50352023 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map