Assay Details
Target Gene Details
Entrez Gene ID: | 1003 |
Gene Name: | cadherin 5 |
Gene Aliases: |
7B4, CD144 |
Location: |
Chr.16:66366607-66404786 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH5 | NM_001795.4 | 3 | 606 | NP_001786.2 |
| XM_011522801.1 | 3 | 583 | XP_011521103.1 | |
| AB035304.1 | 3 | 503 | BAA87418.1 | |
| AB209908.1 | 3 | 506 | BAD93145.1 | |
| AK300299.1 | ||||
| AK300461.1 | ||||
| BC096363.3 | 3 | 496 | AAH96363.1 | |
| BC096364.3 | 3 | 496 | ||
| BC096365.1 | 3 | 496 | AAH96365.1 | |
| BC117520.2 | 3 | 496 | ||
| DC381809.1 | ||||
| U84722.1 | 3 | 554 | AAB41796.1 | |
| X59796.1 | 3 | 526 | CAA42468.1 | |
| X79981.1 | 2 | 459 | CAA56306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833264 | Chr.16:66298692 - 66471069 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map