Assay Details
Target Gene Details
Entrez Gene ID: | 123016 |
Gene Name: | tetratricopeptide repeat domain 8 |
Gene Aliases: |
BBS8, RP51 |
Location: |
Chr.14:88824153-88877996 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC8 | NM_001288781.1 | 2 | 148 | NP_001275710.1 |
| NM_001288782.1 | 2 | 148 | NP_001275711.1 | |
| NM_001288783.1 | 2 | 148 | NP_001275712.1 | |
| NM_144596.3 | 2 | 148 | NP_653197.2 | |
| NM_198309.3 | 2 | 148 | NP_938051.1 | |
| NM_198310.3 | 2 | 148 | NP_938052.1 | |
| XM_006720035.1 | 1 | 430 | XP_006720098.1 | |
| XM_006720037.2 | 1 | 430 | XP_006720100.1 | |
| XM_011536432.1 | 1 | 430 | XP_011534734.1 | |
| XM_011536433.1 | 1 | 430 | XP_011534735.1 | |
| XM_011536434.1 | 1 | 430 | XP_011534736.1 | |
| XM_017020977.1 | 1 | 430 | XP_016876466.1 | |
| XM_017020978.1 | 1 | 430 | XP_016876467.1 | |
| AK093891.1 | ||||
| AK124675.1 | ||||
| AK296077.1 | ||||
| AK315772.1 | ||||
| AL833901.1 | CAD38757.2 | |||
| AY366523.1 | AAR02192.1 | |||
| AY366524.1 | AAR02193.1 | |||
| AY373972.1 | AAR19043.1 | |||
| BC026351.1 | AAH26351.1 | |||
| BC095433.1 | AAH95433.1 | |||
| BX161472.1 | CAD61928.1 | |||
| BX247959.1 | 2 | 148 | CAD62299.1 | |
| BX248071.1 | CAD62360.1 | |||
| KU178849.1 | ||||
| KU178850.1 | ||||
| KU178851.1 | ||||
| KU178852.1 | ||||
| KU178853.1 | ||||
| KU178854.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832850 | Chr.14:88814253 - 88871220 on Build GRCh38 | Gain+Loss |
|
| dgv1955n100 | Chr.14:88712120 - 88999575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map