Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR139 | NM_001002911.3 | 2 | 554 | NP_001002911.1 |
| NM_001318483.1 | 3 | 653 | NP_001305412.1 | |
| AB196529.1 | 2 | 392 | BAD97440.1 | |
| AK291384.1 | 2 | 693 | ||
| AY635179.1 | 2 | 483 | AAT65818.1 | |
| BC137237.1 | 2 | 543 | ||
| BC137238.1 | 2 | 543 | ||
| DN693347.1 | 2 | 554 | ||
| KR081941.1 | 2 | 392 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map