Assay Details
Target Gene Details
Entrez Gene ID: | 1025 |
Gene Name: | cyclin dependent kinase 9 |
Gene Aliases: |
C-2k, CDC2L4, CTK1, PITALRE, TAK |
Location: |
Chr.9:127786026-127790782 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK9 | NM_001261.3 | NP_001252.1 | ||
| XM_017014184.1 | XP_016869673.1 | |||
| AB451289.1 | ||||
| AB451421.1 | ||||
| AK313828.1 | ||||
| BC001968.1 | AAH01968.1 | |||
| BT019903.1 | AAV38706.1 | |||
| L25676.1 | AAA35668.1 | |||
| X80230.1 | CAA56516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12879n54 | Chr.9:127731234 - 127824409 on Build GRCh38 | Loss |
|
| nsv825094 | Chr.9:127742027 - 127802620 on Build GRCh38 | Gain |
|
| nsv471323 | Chr.9:127727464 - 127845239 on Build GRCh38 | Loss |
|
| nsv615373 | Chr.9:127764326 - 127817625 on Build GRCh38 | Loss |
|
| nsv615374 | Chr.9:127770709 - 127866860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map