Assay Details
Target Gene Details
Entrez Gene ID: | 91875 |
Gene Name: | tetratricopeptide repeat domain 5 |
Gene Aliases: |
Strap |
Location: |
Chr.14:20289131-20305994 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC5 | NM_138376.2 | NP_612385.2 | ||
| AK074553.1 | BAC11056.1 | |||
| BC008647.1 | AAH08647.1 | |||
| BC030822.1 | AAH30822.1 | |||
| BC053538.1 | AAH53538.1 | |||
| BX248066.1 | CAD62359.1 | |||
| BX640732.1 | CAE45848.1 | |||
| BX648584.1 | ||||
| DB051711.1 |
More Information
Set Membership: |
|

Genomic Map