Assay Details
Target Gene Details
Entrez Gene ID: | 9499 |
Gene Name: | myotilin |
Gene Aliases: |
LGMD1, LGMD1A, MFM3, TTID, TTOD |
Location: |
Chr.5:137867282-137887851 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOT | NM_001135940.1 | NP_001129412.1 | ||
| NM_001300911.1 | NP_001287840.1 | |||
| NM_006790.2 | NP_006781.1 | |||
| XM_017010060.1 | XP_016865549.1 | |||
| XM_017010061.1 | XP_016865550.1 | |||
| XM_017010062.1 | XP_016865551.1 | |||
| AF133820.2 | AAD29051.2 | |||
| AF144477.1 | AAD44754.1 | |||
| AK300076.1 | ||||
| AK300088.1 | ||||
| AK307620.1 | ||||
| AK312733.1 | ||||
| BC005376.1 | AAH05376.1 | |||
| KU178476.1 | ||||
| KU178477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830501 | Chr.5:137788777 - 137977035 on Build GRCh38 | Gain |
|
| esv3606913 | Chr.5:137856857 - 138124533 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map