Assay Details
Target Gene Details
Entrez Gene ID: | 11128 |
Gene Name: | RNA polymerase III subunit A |
Gene Aliases: |
ADDH, HLD7, RPC1, RPC155, hRPC155 |
Location: |
Chr.10:77975149-78029540 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3A | NM_007055.3 | 31 | 4209 | NP_008986.2 |
| AF021351.1 | 32 | 4079 | AAB86536.1 | |
| AK024744.1 | 3 | 267 | ||
| AK026023.1 | 22 | 2783 | ||
| AK300329.1 | 32 | 4126 | ||
| AY091462.1 | 7 | 803 | AAM12032.1 | |
| BC014399.1 | AAH14399.1 | |||
| BC041089.1 | 31 | 4170 | AAH41089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7310 | Chr.10:77962983 - 78007707 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map