Assay Details
Target Gene Details
Entrez Gene ID: | 5905 |
Gene Name: | Ran GTPase activating protein 1 |
Gene Aliases: |
Fug1, RANGAP, SD |
Location: |
Chr.22:41244777-41302213 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANGAP1 | NM_001278651.1 | 17 | 2996 | NP_001265580.1 |
| NM_001317930.1 | 16 | 2450 | NP_001304859.1 | |
| NM_002883.3 | 16 | 2535 | NP_002874.1 | |
| XM_005261695.1 | 16 | 2663 | XP_005261752.1 | |
| XM_005261696.1 | 16 | 2434 | XP_005261753.1 | |
| XM_006724289.3 | 16 | 2438 | XP_006724352.1 | |
| XM_011530293.1 | 15 | 2528 | XP_011528595.1 | |
| XM_011530294.2 | 16 | 4144 | XP_011528596.1 | |
| XM_011530295.2 | 16 | 2843 | XP_011528597.1 | |
| XM_017028893.1 | 17 | 3774 | XP_016884382.1 | |
| XM_017028894.1 | 17 | 3912 | XP_016884383.1 | |
| XM_017028895.1 | 16 | 4274 | XP_016884384.1 | |
| XM_017028896.1 | 17 | 2662 | XP_016884385.1 | |
| XM_017028897.1 | 16 | 3754 | XP_016884386.1 | |
| XM_017028898.1 | 18 | 3687 | XP_016884387.1 | |
| XM_017028899.1 | 15 | 2400 | XP_016884388.1 | |
| AB058738.1 | 15 | 3719 | ||
| AK303524.1 | 16 | 2458 | ||
| BC004990.1 | 6 | 1110 | AAH04990.1 | |
| BC014044.2 | 16 | 2450 | AAH14044.1 | |
| BC019676.1 | 2 | 651 | AAH19676.1 | |
| BC041396.2 | 17 | 3006 | AAH41396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459888 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| nsv817997 | Chr.22:41244293 - 41366866 on Build GRCh38 | Gain |
|
| nsv589049 | Chr.22:41168704 - 41276267 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map