Assay Details
Target Gene Details
Entrez Gene ID: | 51684 |
Gene Name: | SUFU negative regulator of hedgehog signaling |
Gene Aliases: |
PRO1280, SUFUH, SUFUXL |
Location: |
Chr.10:102502879-102633458 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUFU | NM_001178133.1 | NP_001171604.1 | ||
| NM_016169.3 | NP_057253.2 | |||
| XM_011539858.2 | XP_011538160.1 | |||
| XM_011539860.2 | XP_011538162.1 | |||
| XM_011539861.2 | XP_011538163.1 | |||
| XM_011539863.2 | XP_011538165.1 | |||
| XM_011539864.2 | XP_011538166.1 | |||
| AF144231.1 | AAF23890.1 | |||
| AF159447.1 | AAF23893.1 | |||
| AF172319.1 | AAD51655.1 | |||
| AF175770.1 | AAD50501.1 | |||
| AF222345.1 | AAF35866.1 | |||
| AK312319.1 | ||||
| AY358550.1 | AAQ88914.1 | |||
| BC013291.2 | AAH13291.1 | |||
| DA248208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831968 | Chr.10:102482950 - 102664645 on Build GRCh38 | Gain |
|
| nsv552059 | Chr.10:102501602 - 102627177 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map