Assay Details
Target Gene Details
Entrez Gene ID: | 51399 |
Gene Name: | trafficking protein particle complex 4 |
Gene Aliases: |
CGI-104, HSPC172, PTD009, SBDN, SYNBINDIN, TRS23 |
Location: |
Chr.11:119018469-119023674 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC4 | NM_001318486.1 | NP_001305415.1 | ||
| NM_001318488.1 | NP_001305417.1 | |||
| NM_001318489.1 | NP_001305418.1 | |||
| NM_001318490.1 | NP_001305419.1 | |||
| NM_001318492.1 | NP_001305421.1 | |||
| NM_001318494.1 | NP_001305423.1 | |||
| NM_016146.5 | NP_057230.1 | |||
| AF078862.1 | AAD44494.1 | |||
| AF105025.1 | AAF21897.1 | |||
| AF151862.1 | AAD34099.1 | |||
| AF161520.1 | AAF29135.1 | |||
| AK025623.1 | ||||
| AK290520.1 | ||||
| AK293914.1 | ||||
| AK293977.1 | ||||
| AK297422.1 | ||||
| BC010866.1 | AAH10866.1 | |||
| BF971819.1 | ||||
| BP246594.1 | ||||
| BP332308.1 | ||||
| BU624012.1 |
More Information
Set Membership: |
|

Genomic Map