Assay Details
Target Gene Details
Entrez Gene ID: | 100188893 |
Gene Name: | translocase of outer mitochondrial membrane 6 |
Gene Aliases: |
OBTP, TOM6 |
Location: |
Chr.6:41787443-41789896 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOMM6 | NM_001134493.1 | 1 | 103 | NP_001127965.1 |
| AJ420506.1 | 1 | 812 | ||
| BM783950.1 | 1 | 213 |
More Information
Additional Information:
For this assay, SNP(s) [rs115742810] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map