Assay Details
Target Gene Details
Entrez Gene ID: | 25925 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
EHZF, Evi3 |
Location: |
Chr.18:25061924-25352250 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF521 | NM_001308225.1 | 5 | 3909 | NP_001295154.1 |
| NM_015461.2 | 6 | 3950 | NP_056276.1 | |
| XM_011525909.2 | 6 | 3947 | XP_011524211.1 | |
| XM_011525911.1 | 7 | 3986 | XP_011524213.1 | |
| XM_017025697.1 | 5 | 3836 | XP_016881186.1 | |
| XM_017025698.1 | 7 | 3913 | XP_016881187.1 | |
| AF141339.1 | 3 | 364 | AAG49442.1 | |
| AJ518106.1 | 6 | 3847 | CAD57322.1 | |
| AK021452.1 | 3 | 2014 | BAB13829.1 | |
| AK027354.1 | 6 | 3852 | BAB55056.1 | |
| AK074046.1 | 3 | 3472 | ||
| AK293067.1 | 6 | 3901 | ||
| AL117615.1 | 3 | 1519 | CAB56016.1 | |
| BC032869.2 | 5 | 3808 | AAH32869.2 | |
| BC113622.1 | 6 | 3771 | ||
| BC113648.1 | 6 | 3771 |
More Information
Set Membership: |
|

Genomic Map