Assay Details
Target Gene Details
Entrez Gene ID: | 22868 |
Gene Name: | FAST kinase domains 2 |
Gene Aliases: |
KIAA0971 |
Location: |
Chr.2:206765388-206868284 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FASTKD2 | NM_001136193.1 | 2 | 427 | NP_001129665.1 |
| NM_001136194.1 | 2 | 442 | NP_001129666.1 | |
| NM_014929.3 | 2 | 334 | NP_055744.2 | |
| AB023188.1 | 1 | 75 | BAA76815.2 | |
| AF223470.1 | 2 | 201 | AAF64145.1 | |
| AK001516.1 | 2 | 114 | ||
| AK021557.1 | 2 | 228 | ||
| AL833877.1 | 2 | 108 | CAD38734.1 | |
| BC001544.1 | 1 | 74 | AAH01544.1 | |
| DA021045.1 | 2 | 239 | ||
| DA475994.1 | 2 | 365 | ||
| DA756417.1 | 2 | 224 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011347 | Chr.2:206384007 - 207085773 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map