Assay Details
Target Gene Details
Entrez Gene ID: | 5973 |
Gene Name: | renin binding protein |
Gene Aliases: |
RBP, RNBP |
Location: |
Chr.X:153935263-153944780 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RENBP | NM_002910.5 | NP_002901.2 | ||
| XM_017029698.1 | XP_016885187.1 | |||
| AK298125.1 | ||||
| BC015558.2 | AAH15558.1 | |||
| D10232.1 | BAA01082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map