Assay Details
Target Gene Details
Entrez Gene ID: | 128488 |
Gene Name: | WAP four-disulfide core domain 12 |
Gene Aliases: |
C20orf122, SWAM2, WAP2, dJ211D12.4 |
Location: |
Chr.20:45123426-45124465 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC12 | NM_080869.1 | NP_543145.1 | ||
| AY037803.1 | AAK68848.1 | |||
| AY358678.1 | AAQ89041.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map