Assay Details
Target Gene Details
Entrez Gene ID: | 3145 |
Gene Name: | hydroxymethylbilane synthase |
Gene Aliases: |
PBG-D, PBGD, PORC, UPS |
Location: |
Chr.11:119084871-119093549 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMBS | NM_000190.3 | 14 | 1219 | NP_000181.2 |
| NM_001024382.1 | 14 | 1121 | NP_001019553.1 | |
| NM_001258208.1 | 13 | 1099 | NP_001245137.1 | |
| NM_001258209.1 | 13 | 1190 | NP_001245138.1 | |
| XM_005271531.1 | 14 | 1297 | XP_005271588.1 | |
| XM_005271532.1 | 14 | 1275 | XP_005271589.1 | |
| XM_005271533.3 | 13 | 1171 | XP_005271590.1 | |
| XM_011542796.1 | 13 | 1551 | XP_011541098.1 | |
| XM_017017629.1 | 14 | 1685 | XP_016873118.1 | |
| AK000628.1 | 13 | 1182 | ||
| AK131072.1 | 13 | 1062 | ||
| AK290275.1 | 14 | 1215 | ||
| AW139390.1 | 1 | 308 | ||
| BC000520.2 | 14 | 1213 | AAH00520.1 | |
| BC008149.1 | 14 | 1174 | AAH08149.1 | |
| BC019323.1 | 14 | 1213 | AAH19323.1 | |
| BQ020524.1 | 1 | 309 | ||
| BX647328.1 | 11 | 1824 | ||
| X04217.1 | 14 | 1092 | CAA27801.1 | |
| X04808.1 | 14 | 1089 | CAA28499.1 |
More Information
Set Membership: |
|

Genomic Map