Assay Details
Target Gene Details
Entrez Gene ID: | 91181 |
Gene Name: | nucleoporin 210 like |
Gene Aliases: |
- |
Location: |
Chr.1:153992582-154155116 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP210L | NM_001159484.1 | 34 | 4792 | NP_001152956.1 |
| NM_207308.2 | 34 | 4792 | NP_997191.2 | |
| XM_011510122.1 | 33 | 4623 | XP_011508424.1 | |
| XM_011510123.1 | 33 | 4616 | XP_011508425.1 | |
| XM_011510124.1 | 34 | 4775 | XP_011508426.1 | |
| XM_017002788.1 | 34 | 4775 | XP_016858277.1 | |
| XM_017002789.1 | 34 | 4775 | XP_016858278.1 | |
| AK125924.1 | 30 | 4054 | BAC86345.1 | |
| AK128108.1 | 24 | 3371 | BAC87279.1 | |
| AL133633.1 | 12 | 1566 | CAB63757.1 | |
| AL833964.1 | 18 | 2299 | CAD38812.1 | |
| BX648132.1 | 22 | 2980 |
More Information
Set Membership: |
|

Genomic Map