Assay Details
Target Gene Details
Entrez Gene ID: | 10460 |
Gene Name: | transforming acidic coiled-coil containing protein 3 |
Gene Aliases: |
ERIC-1, ERIC1 |
Location: |
Chr.4:1721383-1745178 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACC3 | NM_006342.2 | 4 | 1058 | NP_006333.1 |
| XM_005247929.2 | 4 | 1056 | XP_005247986.1 | |
| XM_005247930.2 | XP_005247987.1 | |||
| XM_011513386.2 | 4 | 1056 | XP_011511688.1 | |
| XM_017007653.1 | 4 | 1110 | XP_016863142.1 | |
| AF093543.1 | 4 | 1011 | AAD25964.1 | |
| AF479620.1 | AAN76993.1 | |||
| AJ243997.1 | 4 | 1003 | CAB53009.1 | |
| AK225631.1 | 4 | 1026 | ||
| AK302463.1 | 4 | 1029 | ||
| AK304579.1 | 3 | 787 | ||
| AK310660.1 | 3 | 781 | ||
| BC106071.1 | 4 | 1019 | AAI06072.1 | |
| BC111771.1 | 4 | 971 | AAI11772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428434 | Chr.4:1623665 - 1813048 on Build GRCh38 | Gain |
|
| dgv8901n54 | Chr.4:1644637 - 1864631 on Build GRCh38 | Loss |
|
| nsv593316 | Chr.4:1526039 - 1768630 on Build GRCh38 | Gain |
|
| nsv949756 | Chr.4:1710274 - 1817773 on Build GRCh38 | Deletion |
|
| nsv508989 | Chr.4:1623823 - 1846590 on Build GRCh38 | Insertion |
|
| nsv1161027 | Chr.4:1699833 - 1750693 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map