Assay Details
Target Gene Details
Entrez Gene ID: | 57536 |
Gene Name: | KIAA1328 |
Gene Aliases: |
- |
Location: |
Chr.18:36828929-37232127 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1328 | NM_001322327.1 | NP_001309256.1 | ||
| NM_020776.1 | NP_065827.1 | |||
| NR_136303.1 | ||||
| NR_136304.1 | ||||
| NR_136305.1 | ||||
| XM_005258315.4 | XP_005258372.1 | |||
| XM_011526100.2 | XP_011524402.1 | |||
| XM_017025878.1 | XP_016881367.1 | |||
| AK292288.1 | ||||
| AK304230.1 | ||||
| AL832625.2 | CAD89947.1 | |||
| BX648875.1 |
Target Gene Details
Entrez Gene ID: | 25941 |
Gene Name: | tubulin polyglutamylase complex subunit 2 |
Gene Aliases: |
C18orf10, HMFN0601, L17, PGs2 |
Location: |
Chr.18:36777647-36829393 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPGS2 | NM_001271949.1 | NP_001258878.1 | ||
| NM_001271950.1 | NP_001258879.1 | |||
| NM_001271951.1 | NP_001258880.1 | |||
| NM_001271952.1 | NP_001258881.1 | |||
| NM_001271953.1 | NP_001258882.1 | |||
| NM_001271954.1 | NP_001258883.1 | |||
| NM_001271955.1 | NP_001258884.1 | |||
| NM_001271956.1 | NP_001258885.1 | |||
| NM_015476.3 | NP_056291.2 | |||
| XM_005258242.3 | 1 | 205 | XP_005258299.1 | |
| XM_011525917.2 | 1 | 205 | XP_011524219.1 | |
| XM_017025699.1 | 1 | 205 | XP_016881188.1 | |
| XM_017025702.1 | 1 | 205 | XP_016881191.1 | |
| XM_017025703.1 | 1 | 205 | XP_016881192.1 | |
| BC087845.1 | AAH87845.1 | |||
| BG723541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34228 | Chr.18:36536338 - 36849164 on Build GRCh38 | Gain |
|
| esv2758722 | Chr.18:36470072 - 36901904 on Build GRCh38 | Gain |
|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
| nsv438283 | Chr.18:36821197 - 36845836 on Build GRCh38 | Loss |
|
| esv3642237 | Chr.18:36659865 - 36881208 on Build GRCh38 | Gain |
|
| esv3583016 | Chr.18:36337333 - 37122274 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs554635719] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map