Assay Details
Target Gene Details
Entrez Gene ID: | 203427 |
Gene Name: | solute carrier family 25 member 43 |
Gene Aliases: |
- |
Location: |
Chr.X:119399295-119454478 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A43 | NM_145305.2 | 2 | 510 | NP_660348.2 |
| XM_017029340.1 | 2 | 479 | XP_016884829.1 | |
| XM_017029341.1 | 2 | 479 | XP_016884830.1 | |
| AK091304.1 | 2 | 425 | ||
| AK094254.1 | 3 | 325 | BAC04318.1 | |
| AK292035.1 | 2 | 424 | ||
| AK303932.1 | 2 | 425 | ||
| BC019584.1 | 2 | 745 | AAH19584.1 | |
| BC071871.1 | 2 | 472 | AAH71871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518389 | Chr.X:119301240 - 119460459 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map