Assay Details
Target Gene Details
Entrez Gene ID: | 283219 |
Gene Name: | potassium channel tetramerization domain containing 21 |
Gene Aliases: |
KCASH2 |
Location: |
Chr.11:78171243-78188854 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD21 | NM_001029859.2 | 2 | 412 | NP_001025030.1 |
| XM_005273925.3 | 2 | 637 | XP_005273982.1 | |
| XM_005273928.1 | 3 | 537 | XP_005273985.1 | |
| XM_006718516.3 | 2 | 3582 | XP_006718579.1 | |
| XM_006718517.2 | 3 | 649 | XP_006718580.1 | |
| XM_006718518.3 | 2 | 397 | XP_006718581.1 | |
| XM_011544955.2 | 2 | 1877 | XP_011543257.1 | |
| AK095233.1 | ||||
| AK300320.1 | 2 | 399 | ||
| BC036058.1 | 2 | 401 | AAH36058.1 | |
| DC342183.1 | 2 | 412 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss |
|
| nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map