Assay Details
Target Gene Details
Entrez Gene ID: | 85378 |
Gene Name: | tubulin gamma complex associated protein 6 |
Gene Aliases: |
GCP-6, GCP6, MCCRP, MCCRP1, MCPHCR |
Location: |
Chr.22:50217689-50245024 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP6 | NM_020461.3 | 16 | 3452 | NP_065194.2 |
| AB051456.1 | 16 | 2846 | BAB33339.1 | |
| AF272887.1 | 16 | 3045 | AAK82968.1 | |
| AL137665.1 | 9 | 1269 | ||
| AL832617.2 | 16 | 3473 | ||
| BC023573.2 | 2 | 512 | AAH23573.1 | |
| BC144488.1 | 16 | 3020 | ||
| BC150613.1 | 16 | 3020 | ||
| BC151209.1 | 16 | 3434 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map