Assay Details
Target Gene Details
Entrez Gene ID: | 51409 |
Gene Name: | HemK methyltransferase family member 1 |
Gene Aliases: |
HEMK, MTQ1 |
Location: |
Chr.3:50569152-50589050 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEMK1 | NM_001317851.1 | 11 | 2974 | NP_001304780.1 |
| NM_016173.4 | 11 | 3275 | NP_057257.1 | |
| XM_005265218.3 | 9 | 3119 | XP_005265275.1 | |
| XM_011533806.2 | 11 | 2044 | XP_011532108.1 | |
| XM_011533807.1 | 11 | 1929 | XP_011532109.1 | |
| XM_011533808.1 | 11 | 1715 | XP_011532110.1 | |
| XM_011533809.2 | XP_011532111.1 | |||
| XM_011533810.2 | XP_011532112.1 | |||
| XM_011533812.2 | 9 | 1888 | XP_011532114.1 | |
| XM_011533813.2 | 11 | 2082 | XP_011532115.1 | |
| XM_011533814.2 | 11 | 2006 | XP_011532116.1 | |
| AF172244.1 | 11 | 2875 | AAD51328.1 | |
| BX537562.1 | 10 | 3087 | ||
| DA404565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584502 | Chr.3:50327055 - 50622310 on Build GRCh38 | Gain |
|
| nsv590273 | Chr.3:50536782 - 50640720 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map