Assay Details
Target Gene Details
Entrez Gene ID: | 80758 |
Gene Name: | proline rich 7 (synaptic) |
Gene Aliases: |
- |
Location: |
Chr.5:177445936-177456701 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR7 | NM_001174101.1 | 2 | 427 | NP_001167572.1 |
| NM_001174102.1 | 2 | 344 | NP_001167573.1 | |
| NM_030567.4 | 3 | 512 | NP_085044.2 | |
| XM_011534662.1 | 2 | 6250 | XP_011532964.1 | |
| XM_011534663.2 | 2 | 988 | XP_011532965.1 | |
| XM_017009896.1 | 1 | 7038 | XP_016865385.1 | |
| XM_017009897.1 | 2 | 1220 | XP_016865386.1 | |
| BC004261.1 | 2 | 407 | AAH04261.1 | |
| BC021240.1 | 2 | 335 | AAH21240.1 | |
| BC024233.2 | 3 | 516 | AAH24233.2 | |
| BC033649.1 | 1 | 102 | AAH33649.1 | |
| BE277571.1 | 2 | 376 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161313 | Chr.5:177356552 - 177463415 on Build GRCh38 | Duplication |
|
| dgv733n27 | Chr.5:177357511 - 177469411 on Build GRCh38 | Loss |
|
| nsv515805 | Chr.5:177432847 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|

Genomic Map