Assay Details
Target Gene Details
Entrez Gene ID: | 5128 |
Gene Name: | cyclin dependent kinase 17 |
Gene Aliases: |
PCTAIRE2, PCTK2 |
Location: |
Chr.12:96278261-96400588 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK17 | NM_001170464.2 | 16 | 4521 | NP_001163935.1 |
| NM_002595.4 | 17 | 3993 | NP_002586.2 | |
| AF086067.1 | 1 | 274 | ||
| BF111798.1 | 1 | 211 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv836 | Chr.12:96259232 - 96293939 on Build GRCh38 | Insertion |
|
| nsv1035411 | Chr.12:96237097 - 96338527 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map