Assay Details
Target Gene Details
Entrez Gene ID: | 10103 |
Gene Name: | tetraspanin 1 |
Gene Aliases: |
NET1, TM4C, TM4SF |
Location: |
Chr.1:46175077-46196489 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN1 | NM_005727.3 | 9 | 1249 | NP_005718.2 |
| XM_011540460.2 | XP_011538762.1 | |||
| AF054838.1 | 7 | 775 | AAC69714.1 | |
| AF065388.1 | 8 | 896 | AAC17119.1 | |
| AF133425.1 | 8 | 898 | AAF08364.1 | |
| BC007290.1 | 8 | 902 | AAH07290.1 | |
| BC013404.2 | 8 | 900 | AAH13404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| nsv527878 | Chr.1:46183374 - 46194623 on Build GRCh38 | Gain |
|
| nsv822520 | Chr.1:46184814 - 46196029 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map