Assay Details
Target Gene Details
Entrez Gene ID: | 55216 |
Gene Name: | chromosome 11 open reading frame 57 |
Gene Aliases: |
- |
Location: |
Chr.11:112074244-112085150 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf57 | NM_001082969.1 | 4 | 365 | NP_001076438.1 |
| NM_001082970.1 | 4 | 923 | NP_001076439.1 | |
| NM_001301017.1 | 4 | 365 | NP_001287946.1 | |
| NM_001301019.1 | 4 | 406 | NP_001287948.1 | |
| NM_001301021.1 | 4 | 406 | NP_001287950.1 | |
| NM_018195.3 | 4 | 923 | NP_060665.3 | |
| NR_103469.1 | 4 | 964 | ||
| AB096251.1 | 4 | 269 | ||
| AK001588.1 | BAA91774.1 | |||
| AK094203.1 | 4 | 299 | ||
| AK125340.1 | 4 | 879 | BAC86137.1 | |
| BC005403.1 | 2 | 162 | ||
| BC030546.1 | 2 | 167 | ||
| BC048133.1 | 4 | 851 | ||
| BC072455.1 | 4 | 290 | ||
| BG115155.1 | 4 | 413 | ||
| BX538107.1 | 4 | 922 | CAD98020.1 | |
| CD244552.1 | 4 | 324 | ||
| DA813095.1 | 4 | 351 | ||
| EL736728.1 | 2 | 155 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map