Assay Details
Target Gene Details
Entrez Gene ID: | 84000 |
Gene Name: | transmembrane protease, serine 13 |
Gene Aliases: |
MSP, MSPL, MSPS, TMPRSS11 |
Location: |
Chr.11:117900641-117929453 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS13 | NM_001077263.2 | 13 | 2829 | NP_001070731.1 |
| NM_001206789.1 | 12 | 2724 | NP_001193718.1 | |
| AB048797.1 | 12 | 2680 | ||
| BC114928.1 | 13 | 2748 | AAI14929.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74491696,rs78306421] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map