Assay Details
Target Gene Details
Entrez Gene ID: | 5360 |
Gene Name: | phospholipid transfer protein |
Gene Aliases: |
BPIFE, HDLCQ9 |
Location: |
Chr.20:45898620-45912364 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLTP | NM_001242920.1 | NP_001229849.1 | ||
| NM_001242921.1 | NP_001229850.1 | |||
| NM_006227.3 | NP_006218.1 | |||
| NM_182676.2 | NP_872617.1 | |||
| AB076694.1 | BAB79630.1 | |||
| AF052157.1 | ||||
| AK092397.1 | ||||
| AK092523.1 | ||||
| AK097009.1 | ||||
| AK222690.1 | BAD96410.1 | |||
| AK289371.1 | ||||
| AK293150.1 | ||||
| AK299181.1 | ||||
| BC005045.1 | AAH05045.1 | |||
| BC019847.1 | AAH19847.1 | |||
| BC019898.1 | AAH19898.1 | |||
| L26232.1 | AAA36443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517296 | Chr.20:45854586 - 45965936 on Build GRCh38 | Loss |
|
| nsv833991 | Chr.20:45885631 - 46068708 on Build GRCh38 | Loss |
|
| nsv586134 | Chr.20:45899835 - 45905120 on Build GRCh38 | Gain |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map