Assay Details
Target Gene Details
Entrez Gene ID: | 33 |
Gene Name: | acyl-CoA dehydrogenase, long chain |
Gene Aliases: |
ACAD4, LCAD |
Location: |
Chr.2:210187990-210225491 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACADL | NM_001608.3 | 11 | 1487 | NP_001599.1 |
| XM_005246517.4 | 11 | 1939 | XP_005246574.1 | |
| XM_017003955.1 | 11 | 1915 | XP_016859444.1 | |
| AK313498.1 | ||||
| BC039063.1 | 11 | 1432 | AAH39063.1 | |
| BC064549.1 | 11 | 1419 | AAH64549.1 | |
| M74096.1 | 11 | 1264 | AAA51565.1 |
More Information
Set Membership: |
|

Genomic Map