Assay Details
Target Gene Details
Entrez Gene ID: | 710 |
Gene Name: | serpin family G member 1 |
Gene Aliases: |
C1IN, C1INH, C1NH, HAE1, HAE2 |
Location: |
Chr.11:57597554-57614853 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPING1 | NM_000062.2 | NP_000053.2 | ||
| AL703442.1 | 1 | 66 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
|
| nsv1126294 | Chr.11:57597027 - 57598327 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map