Assay Details
Target Gene Details
Entrez Gene ID: | 11217 |
Gene Name: | A-kinase anchoring protein 2 |
Gene Aliases: |
AKAP-2, AKAPKL, MISP2, PRKA2 |
Location: |
Chr.9:110048598-110172512 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP2 | NM_001004065.4 | NP_001004065.2 | ||
| NM_001136562.2 | NP_001130034.1 | |||
| NM_001198656.1 | NP_001185585.1 | |||
| AK000306.1 | ||||
| AK022693.1 | ||||
| AK057098.1 | ||||
| AK300427.1 | ||||
| AK304311.1 | ||||
| AL110268.1 | CAB53707.1 | |||
| BC146863.1 | ||||
| BC171800.1 |
Target Gene Details
Entrez Gene ID: | 445815 |
Gene Name: | PALM2-AKAP2 readthrough |
Gene Aliases: |
AKAP2 |
Location: |
Chr.9:109780297-110172512 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALM2-AKAP2 | NM_007203.4 | NP_009134.1 | ||
| NM_147150.2 | NP_671492.1 | |||
| AB023137.1 | ||||
| AJ303079.1 | CAC38839.1 | |||
| BC140818.1 | ||||
| BX649126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv494n21 | Chr.9:109952819 - 110202456 on Build GRCh38 | Gain |
|
| esv3621449 | Chr.9:109959589 - 110304023 on Build GRCh38 | Gain |
|
| esv3621453 | Chr.9:110129325 - 110304387 on Build GRCh38 | Gain |
|
| nsv615151 | Chr.9:110157008 - 110175898 on Build GRCh38 | Gain |
|
| nsv1040651 | Chr.9:110161896 - 110308298 on Build GRCh38 | Gain |
|
| nsv515562 | Chr.9:110157886 - 110175898 on Build GRCh38 | Gain |
|
| nsv1043494 | Chr.9:109988131 - 110214627 on Build GRCh38 | Gain |
|
| esv3621456 | Chr.9:110153125 - 110182880 on Build GRCh38 | Gain |
|

Genomic Map