Assay Details
Target Gene Details
Entrez Gene ID: | 84340 |
Gene Name: | G elongation factor mitochondrial 2 |
Gene Aliases: |
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2 |
Location: |
Chr.5:74721204-74767371 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFM2 | NM_001281302.1 | NP_001268231.1 | ||
| NM_032380.4 | NP_115756.2 | |||
| NM_170681.2 | NP_733781.1 | |||
| NM_170691.2 | NP_733792.1 | |||
| NR_104006.1 | ||||
| XM_011543691.2 | XP_011541993.1 | |||
| XM_017009986.1 | XP_016865475.1 | |||
| AF111808.1 | AAL39010.1 | |||
| AF367997.1 | AAK53401.1 | |||
| AK025314.1 | BAB15109.1 | |||
| AK307839.1 | ||||
| AK308422.1 | ||||
| AK315705.1 | ||||
| AY453587.1 | ||||
| BC015712.1 | AAH15712.1 | |||
| BC030612.1 | AAH30612.1 | |||
| BI828937.1 | ||||
| DC402212.1 |
Target Gene Details
Entrez Gene ID: | 10412 |
Gene Name: | NSA2, ribosome biogenesis homolog |
Gene Aliases: |
CDK105, HCL-G1, HCLG1, HUSSY-29, HUSSY29, TINP1 |
Location: |
Chr.5:74765429-74776912 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSA2 | NM_001271665.1 | NP_001258594.1 | ||
| NM_014886.4 | NP_055701.1 | |||
| NR_073403.1 | ||||
| XM_011543098.1 | XP_011541400.1 | |||
| XM_017008952.1 | XP_016864441.1 | |||
| HY021659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598653 | Chr.5:74697056 - 74799717 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map