Assay Details
Target Gene Details
Entrez Gene ID: | 80010 |
Gene Name: | RecQ mediated genome instability 1 |
Gene Aliases: |
BLAP75, C9orf76, FAAP75 |
Location: |
Chr.9:83980722-84004074 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMI1 | NM_024945.2 | 1 | 138 | NP_079221.2 |
| XM_005252211.2 | XP_005252268.1 | |||
| XM_005252213.2 | XP_005252270.1 | |||
| XM_011519034.2 | 1 | 120 | XP_011517336.1 | |
| AK022950.1 | 1 | 62 | BAB14325.1 | |
| BC020606.1 | ||||
| BC032494.1 | ||||
| BC039999.2 | AAH39999.1 | |||
| BC053549.1 | AAH53549.1 | |||
| BP327553.1 | ||||
| DN997105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831644 | Chr.9:83919395 - 84089480 on Build GRCh38 | Gain |
|
| esv2759701 | Chr.9:83799109 - 83988687 on Build GRCh38 | Gain |
|
| nsv1043703 | Chr.9:83931002 - 83988630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map