Assay Details
Target Gene Details
Entrez Gene ID: | 5594 |
Gene Name: | mitogen-activated protein kinase 1 |
Gene Aliases: |
ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK |
Location: |
Chr.22:21759657-21867680 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK1 | NM_002745.4 | 9 | 5277 | NP_002736.3 |
| AK055080.1 | ||||
| BC099905.1 | AAH99905.1 | |||
| EF565108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588322 | Chr.22:21732084 - 21832190 on Build GRCh38 | Loss |
|
| nsv510804 | Chr.22:21670242 - 21778475 on Build GRCh38 | Deletion |
|
| nsv1060548 | Chr.22:21724874 - 21772537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map