Assay Details
Target Gene Details
Entrez Gene ID: | 79918 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.16:58515479-58521961 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD6 | NM_001160305.2 | 4 | 587 | NP_001153777.1 |
| NM_024860.3 | 5 | 515 | NP_079136.2 | |
| NR_134583.1 | 5 | 515 | ||
| XM_011523336.1 | 4 | 578 | XP_011521638.1 | |
| AK024801.1 | 5 | 471 | BAB15011.1 | |
| AK127136.1 | 4 | 564 | BAC86845.1 | |
| AK290495.1 | 4 | 549 | ||
| AK297288.1 | 4 | 690 | ||
| AK304822.1 | 4 | 628 | ||
| AK309468.1 | 4 | 546 | ||
| BC022451.1 | 4 | 549 | AAH22451.1 | |
| BP338820.1 | 5 | 514 |
More Information
Set Membership: |
|

Genomic Map