Assay Details
Target Gene Details
Entrez Gene ID: | 5375 |
Gene Name: | peripheral myelin protein 2 |
Gene Aliases: |
FABP8, M-FABP, MP2, P2 |
Location: |
Chr.8:81440326-81447484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMP2 | NM_002677.3 | NP_002668.1 | ||
| AK307118.1 | ||||
| AK311758.1 | ||||
| BC034997.1 | AAH34997.1 | |||
| CR541649.1 | CAG46450.1 | |||
| CR541738.1 | CAG46538.1 | |||
| X62167.1 | CAA44096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465721 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map