Assay Details
Target Gene Details
Entrez Gene ID: | 514 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit |
Gene Aliases: |
ATPE, MC5DN3 |
Location: |
Chr.20:59028678-59032367 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5E | NM_006886.3 | NP_008817.1 | ||
| AF052955.1 | AAF72736.1 | |||
| AF077045.1 | AAD27778.1 | |||
| AK026556.1 | ||||
| BC001690.1 | AAH01690.1 | |||
| BC003671.1 | AAH03671.1 | |||
| BC105811.1 | AAI05812.1 | |||
| CB106920.1 | ||||
| DA202623.1 |
Target Gene Details
Entrez Gene ID: | 100533975 |
Gene Name: | SLMO2-ATP5E readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:59028678-59042846 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLMO2-ATP5E | NR_037929.1 | |||
| NR_037930.1 | ||||
| CD522026.1 | ||||
| FY212904.1 |
More Information
Set Membership: |
|

Genomic Map