Assay Details
Target Gene Details
Entrez Gene ID: | 51510 |
Gene Name: | charged multivesicular body protein 5 |
Gene Aliases: |
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60 |
Location: |
Chr.9:33264879-33282069 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP5 | NM_001195536.1 | 1 | 169 | NP_001182465.1 |
| NM_016410.5 | 1 | 169 | NP_057494.3 | |
| AF132968.1 | AAD27743.1 | |||
| AF161525.1 | AAF29140.1 | |||
| AK024753.1 | ||||
| AK295744.1 | ||||
| AK315455.1 | ||||
| BC006974.1 | AAH06974.1 | |||
| BC007457.1 | AAH07457.1 | |||
| BC016698.1 | 1 | 48 | AAH16698.1 | |
| BC020796.1 | AAH20796.1 | |||
| BC021168.1 | AAH21168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519369 | Chr.9:33134619 - 33283163 on Build GRCh38 | Gain |
|
| dgv7516n100 | Chr.9:33121909 - 33278846 on Build GRCh38 | Gain |
|
| nsv614146 | Chr.9:33186158 - 33398608 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map